Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026512.1 | NP_080788.1 | 7 | 846 | ||
GenBank mRNA | AK002907.1 | - | 7 | 844 | ||
AK007606.1 | - | 7 | 841 | |||
AK008202.1 | - | 7 | 865 | |||
AK078232.1 | - | 7 | 891 | |||
AK152712.1 | - | 7 | 856 | |||
AK170099.1 | - | 7 | 849 | |||
BC025162.1 | - | 7 | 846 | 4921451 | ||
BC029322.1 | - | 5 | 481 |