Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026512.1 | NP_080788.1 | 4 | 478 | ||
GenBank mRNA | AK002907.1 | - | 4 | 476 | ||
AK007606.1 | - | 4 | 472 | |||
AK008202.1 | - | 4 | 496 | |||
AK017801.1 | - | 4 | 481 | |||
AK078232.1 | - | 4 | 523 | |||
AK152712.1 | - | 4 | 488 | |||
AK170099.1 | - | 4 | 481 | |||
BC023146.1 | - | 4 | 463 | 3990118 | ||
BC025162.1 | - | 4 | 478 | 4921451 | ||
BC029322.1 | - | 2 | 113 |