Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810417H13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026515.2 | NP_080791.2 | 2 | 204 | ||
GenBank mRNA | AK011090.1 | - | 2 | 210 | ||
AK011645.1 | - | 2 | 167 | |||
AK011835.1 | - | 2 | 164 | |||
AK012140.1 | - | 2 | 174 | |||
AK013105.1 | - | 2 | 164 | |||
AK017673.1 | - | 2 | 167 | |||
AK134432.1 | - | 2 | 167 | |||
AK212319.1 | - | 2 | 76 | |||
AY496943.1 | - | 2 | 98 | |||
BC021413.1 | - | 2 | 150 | 5006272 | ||
BC096393.1 | - | 2 | 140 | 1397865 |