Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810417H13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026515.2 | NP_080791.2 | 3 | 344 | ||
GenBank mRNA | AK011090.1 | - | 3 | 350 | ||
AK011645.1 | - | 3 | 307 | |||
AK011835.1 | - | 3 | 304 | |||
AK012140.1 | - | 3 | 314 | |||
AK013105.1 | - | 3 | 304 | |||
AK017673.1 | - | 3 | 307 | |||
AY496943.1 | - | 3 | 238 | |||
BC021413.1 | - | 3 | 290 | 5006272 | ||
BC096393.1 | - | 3 | 280 | 1397865 |