Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl22l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026517.2 | NP_080793.1 | 4 | 278 | ||
GenBank mRNA | AK008915.1 | - | 4 | 280 | ||
AK010265.1 | - | 4 | 258 | |||
AK013966.1 | - | 4 | 278 | |||
AK170359.1 | - | 1 | 1710 | |||
BC026533.1 | - | 4 | 258 | 5030430 | ||
BC120716.1 | - | 4 | 353 | 40129639 | ||
BC120718.1 | - | 4 | 353 | 40129641 |