Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133693.2 | NP_598454.2 | 2, 3 | 425 | ||
GenBank mRNA | AK013929.1 | - | 2, 3 | 363 | ||
AK151000.1 | - | 2, 3 | 337 | |||
AK194325.1 | - | 1, 2 | 31 | |||
AK197356.1 | - | 1, 2 | 31 | |||
AK210537.1 | - | 1, 2 | 31 | |||
AK211257.1 | - | 1, 2 | 78 | |||
AK216265.1 | - | 1, 2 | 78 | |||
BC009148.1 | - | 2, 3 | 362 | 3600406 | ||
BC011286.1 | - | 2, 3 | 344 | 3485410 | ||
BC027372.1 | - | 2, 3 | 349 | 4952304 | ||
BC039917.1 | - | 2, 3 | 360 | |||
BC057928.1 | - | 2, 3 | 351 | 5289543 |