Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026521.4 | NP_080797.1 | 4 | 729 | ||
GenBank mRNA | AB041652.1 | - | 4 | 672 | ||
AK004076.1 | - | 4 | 528 | |||
AK014015.1 | - | 1 | 70 | |||
AK032231.1 | - | 4 | 615 | |||
AK082392.1 | - | 4 | 633 | |||
AK132053.1 | - | 4 | 646 | |||
AK145319.1 | - | 4 | 607 | |||
AK166530.1 | - | 4 | 607 | |||
AK167791.1 | - | 4 | 718 | |||
AK167897.1 | - | 4 | 646 | |||
AK168303.1 | - | 4 | 646 | |||
AK168958.1 | - | 4 | 608 | |||
AK197011.1 | - | 1 | 155 | |||
AK200254.1 | - | 1 | 155 | |||
BC037600.1 | - | 4 | 654 | |||
BC042704.1 | - | 4 | 620 | |||
BC052459.1 | - | 4 | 729 | |||
BC054356.1 | - | 4 | 651 | |||
BC085477.1 | - | 4 | 613 | |||
BC147306.1 | - | 4 | 588 | 9056379 | ||
BC147307.1 | - | 4 | 588 | 9056367 |