Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026521.4 | NP_080797.1 | 2 | 385 | ||
GenBank mRNA | AB041652.1 | - | 2 | 328 | ||
AK004076.1 | - | 2 | 184 | |||
AK032231.1 | - | 2 | 271 | |||
AK082392.1 | - | 2 | 289 | |||
AK132053.1 | - | 2 | 302 | |||
AK145319.1 | - | 2 | 263 | |||
AK166530.1 | - | 2 | 263 | |||
AK167791.1 | - | 2 | 373 | |||
AK167897.1 | - | 2 | 302 | |||
AK168303.1 | - | 2 | 302 | |||
AK168958.1 | - | 2 | 264 | |||
AK187810.1 | - | 1 | 113 | |||
BC037600.1 | - | 2 | 310 | |||
BC042704.1 | - | 2 | 276 | |||
BC052459.1 | - | 2 | 385 | |||
BC054356.1 | - | 2 | 307 | |||
BC085477.1 | - | 2 | 269 | |||
BC147306.1 | - | 2 | 244 | 9056379 | ||
BC147307.1 | - | 2 | 244 | 9056367 |