Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142681.1 | NP_001136153.1 | 4 | 459 | ||
NM_026522.5 | NP_080798.2 | 4 | 459 | |||
GenBank mRNA | AK014071.1 | - | 4 | 408 | ||
AK014141.1 | - | 4 | 434 | |||
AK036717.1 | - | 4 | 403 | |||
AK050655.1 | - | 4 | 407 | |||
AK147377.1 | - | 4 | 422 | |||
AK160156.1 | - | 4 | 419 | |||
AK160200.1 | - | 4 | 419 | |||
BC006876.1 | - | 4 | 408 | 2647540 | ||
BC061063.1 | - | 4 | 419 | 6591338 |