Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012328.2 | NP_036460.1 | 3 | 989 | ||
GenBank mRNA | AB026908.1 | - | 3 | 822 | ||
AF083247.1 | - | 3 | 823 | |||
AF093824.1 | - | 3 | 789 | |||
AK092204.1 | - | 4 | 847 | |||
AL080081.1 | - | 3 | 869 | |||
AY359045.1 | - | 2 | 643 | |||
BC028912.1 | - | 3 | 803 | 4943933 | ||
CR533475.1 | - | 2 | 620 |