Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026542.3 | NP_080818.1 | 9 | 882 | ||
GenBank mRNA | AK002381.1 | - | 9 | 838 | ||
AK007276.1 | - | 9 | 861 | |||
AK007934.1 | - | 9 | 836 | |||
AK010272.1 | - | 9 | 859 | |||
AK013834.1 | - | 5 | 526 | |||
AK019396.1 | - | 9 | 874 | |||
AK078417.1 | - | 9 | 874 | |||
AK084116.1 | - | 9 | 873 | |||
AK160922.1 | - | 9 | 828 | |||
AK161289.1 | - | 9 | 874 | |||
AK211485.1 | - | 4 | 365 | |||
BC115661.1 | - | 9 | 782 | 40103267 | ||
BC115662.1 | - | 9 | 789 | 40103268 |