Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026542.3 | NP_080818.1 | 12 | 1162 | ||
GenBank mRNA | AK002381.1 | - | 12 | 1117 | ||
AK007276.1 | - | 12 | 1140 | |||
AK007934.1 | - | 12 | 1116 | |||
AK010272.1 | - | 12 | 1138 | |||
AK013834.1 | - | 8 | 806 | |||
AK019396.1 | - | 12 | 1154 | |||
AK078417.1 | - | 12 | 1154 | |||
AK084116.1 | - | 11 | 1230 | |||
AK160922.1 | - | 12 | 1108 | |||
AK161289.1 | - | 12 | 1154 | |||
BC115661.1 | - | 12 | 1062 | 40103267 | ||
BC115662.1 | - | 12 | 1069 | 40103268 |