Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199111.1 | NP_001186040.1 | 8 | 1099 | ||
NM_001199112.1 | NP_001186041.1 | 7 | 1002 | |||
NM_001316374.1 | NP_001303303.1 | 8 | 1303 | |||
NM_005917.3 | NP_005908.1 | 8 | 1303 | |||
GenBank mRNA | AK295931.1 | - | 8 | 1017 | ||
AK300719.1 | - | 7 | 828 | |||
AK312331.1 | - | 8 | 944 | |||
BC001484.2 | - | 8 | 924 | 3505345 | ||
CR457405.1 | - | 7 | 868 | |||
D55654.1 | - | 8 | 923 | |||
EU794635.1 | - | 8 | 924 | |||
U20352.1 | - | 7 | 868 |