Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170486.1 | NP_001163957.1 | 2 | 156 | ||
NM_026552.3 | NP_080828.1 | 3 | 275 | |||
GenBank mRNA | AJ278129.1 | - | 3 | 201 | ||
AK017993.1 | - | 3 | 276 | |||
AK018069.1 | - | 3 | 213 | |||
AK030840.1 | - | 1 | 3858 | |||
AK039215.1 | - | 3 | 223 | |||
AK076248.1 | - | 3 | 223 | |||
AK077325.1 | - | 3 | 197 | |||
AK133626.1 | - | 3 | 223 | |||
AK136082.1 | - | 3 | 218 | |||
AK150690.1 | - | 3 | 209 | |||
AK153380.1 | - | 3 | 207 | |||
AK159410.1 | - | 2 | 104 | |||
AK166440.1 | - | 3 | 197 | |||
AK193553.1 | - | 2 | 146 | |||
AK194624.1 | - | 2 | 146 | |||
AK194784.1 | - | 2 | 146 | |||
BC015280.1 | - | 2 | 187 | |||
BC055309.1 | - | 2 | 186 | 6529255 |