Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170486.1 | NP_001163957.1 | 2 | 110 | ||
NM_026552.3 | NP_080828.1 | 3 | 229 | |||
GenBank mRNA | AJ278129.1 | - | 3 | 155 | ||
AK017993.1 | - | 3 | 230 | |||
AK018069.1 | - | 3 | 167 | |||
AK030840.1 | - | 1 | 3812 | |||
AK039215.1 | - | 3 | 177 | |||
AK076248.1 | - | 3 | 177 | |||
AK077325.1 | - | 3 | 151 | |||
AK133626.1 | - | 3 | 177 | |||
AK136082.1 | - | 3 | 172 | |||
AK150690.1 | - | 3 | 163 | |||
AK153380.1 | - | 3 | 161 | |||
AK159410.1 | - | 2 | 58 | |||
AK166440.1 | - | 3 | 151 | |||
AK193553.1 | - | 2 | 100 | |||
AK194784.1 | - | 2 | 100 | |||
AK197517.1 | - | 2 | 100 | |||
BC015280.1 | - | 2 | 141 | |||
BC055309.1 | - | 2 | 140 | 6529255 |