Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170485.1 | NP_001163956.1 | 2, 3 | 187 | ||
NM_001170486.1 | NP_001163957.1 | 3, 4 | 299 | |||
NM_026552.3 | NP_080828.1 | 4, 5 | 418 | |||
GenBank mRNA | AJ278129.1 | - | 4, 5 | 344 | ||
AK017993.1 | - | 4, 5 | 419 | |||
AK018069.1 | - | 4, 5 | 356 | |||
AK039215.1 | - | 4, 5 | 366 | |||
AK076248.1 | - | 4, 5 | 366 | |||
AK077325.1 | - | 4, 5 | 340 | |||
AK133626.1 | - | 4, 5 | 366 | |||
AK136082.1 | - | 4, 5 | 361 | |||
AK150690.1 | - | 4, 5 | 352 | |||
AK153380.1 | - | 4, 5 | 350 | |||
AK159410.1 | - | 3, 4 | 247 | |||
AK166440.1 | - | 4, 5 | 340 | |||
BC015280.1 | - | 3, 4 | 330 | |||
BC055309.1 | - | 3, 4 | 329 | 6529255 |