Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rchy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271797.1 | NP_001258726.1 | 7 | 806 | ||
NM_026557.4 | NP_080833.1 | 8 | 926 | |||
GenBank mRNA | AF071222.1 | - | 8 | 598 | ||
AF276959.1 | - | 8 | 596 | |||
AF305423.1 | - | 8 | 582 | |||
AK015870.1 | - | 9 | 919 | |||
AK018364.1 | - | 8 | 593 | |||
AK018488.1 | - | 8 | 623 | |||
AK078397.1 | - | 8 | 710 | |||
BC023138.1 | - | 8 | 700 | 4008685 | ||
BC057143.1 | - | 8 | 926 | 5249543 | ||
DV662964.1 | - | 7 | 473 |