Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rchy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271797.1 | NP_001258726.1 | 3 | 617 | ||
NM_026557.4 | NP_080833.1 | 4 | 737 | |||
GenBank mRNA | AF071222.1 | - | 4 | 409 | ||
AF276959.1 | - | 4 | 407 | |||
AF305423.1 | - | 4 | 393 | |||
AK015870.1 | - | 5 | 730 | |||
AK018364.1 | - | 4 | 404 | |||
AK018488.1 | - | 4 | 434 | |||
AK078397.1 | - | 4 | 521 | |||
BC023138.1 | - | 4 | 511 | 4008685 | ||
BC057143.1 | - | 4 | 737 | 5249543 | ||
DV662964.1 | - | 3 | 284 |