Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282403.1 | NP_001269332.1 | 2, 3 | 371 | ||
NM_005918.3 | NP_005909.2 | 2, 3 | 371 | |||
GenBank mRNA | AF047470.1 | - | 2, 3 | 315 | ||
AK290779.1 | - | 2, 3 | 305 | |||
AK293460.1 | - | 2, 3 | 229 | |||
AK316587.1 | - | 2, 3 | 254 | |||
BC001917.1 | - | 2, 3 | 257 | 2823443 | ||
BP382246.1 | - | 2, 3 | 448 | |||
CR536548.1 | - | 2, 3 | 229 | |||
DC354622.1 | - | 2, 3 | 348 | |||
DQ402957.1 | - | 1, 2 | 166 |