Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282403.1 | NP_001269332.1 | 3 | 420 | ||
NM_001282404.1 | NP_001269333.1 | 2 | 251 | |||
NM_005918.3 | NP_005909.2 | 3 | 420 | |||
GenBank mRNA | AF047470.1 | - | 3 | 364 | ||
AK095803.1 | - | 2 | 185 | |||
AK290779.1 | - | 3 | 354 | |||
AK293460.1 | - | 3 | 278 | |||
AK316587.1 | - | 3 | 303 | |||
BC001917.1 | - | 3 | 306 | 2823443 | ||
BP382246.1 | - | 3 | 497 | |||
CR536548.1 | - | 3 | 278 | |||
DC354622.1 | - | 3 | 397 | |||
DQ402957.1 | - | 2 | 215 |