Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023431.5 | NP_075920.4 | 10 | 1554 | ||
GenBank mRNA | AK011465.1 | - | 1 | 58 | ||
AK078792.1 | - | 7 | 1261 | |||
AK147490.1 | - | 2 | 3992 | |||
AK154610.1 | - | 9 | 1941 | |||
AK158750.1 | - | 10 | 1554 | |||
AK159972.1 | - | 9 | 2456 | |||
AK170459.1 | - | 10 | 1473 | |||
AK178196.1 | - | 3 | 175 | |||
AK186340.1 | - | 2 | 390 | |||
AK192032.1 | - | 5 | 297 | |||
AK219139.1 | - | 3 | 175 | |||
BC008550.1 | - | 7 | 1227 | |||
BC023031.1 | - | 5 | 312 | |||
BC057192.1 | - | 3 | 2927 | |||
BC069883.1 | - | 3 | 190 | |||
BC075617.1 | - | 9 | 1443 | 5718202 |