Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024217.3 | NP_077179.1 | 4 | 593 | ||
GenBank mRNA | AK003882.1 | - | 4 | 449 | ||
AK172091.1 | - | 4 | 496 | |||
AY241870.1 | - | 4 | 587 | |||
BC003230.1 | - | 4 | 500 | 3483351 | ||
BC145817.1 | - | 4 | 465 | 40131120 | ||
BC145819.1 | - | 4 | 465 | 40131122 |