Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026573.2 | NP_080849.1 | 4, 5 | 557 | ||
GenBank mRNA | AK011606.1 | - | 4, 5 | 555 | ||
AK019989.1 | - | 4, 5 | 535 | |||
AK145431.1 | - | 4, 5 | 560 | |||
AK162222.1 | - | 4, 5 | 553 | |||
BC047049.1 | - | 2, 3 | 216 | |||
BC058761.1 | - | 4, 5 | 538 |