Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026576.3 | NP_080852.2 | 5 | 1345 | ||
GenBank mRNA | AK018594.1 | - | 1 | 642 | ||
AK019965.1 | - | 5 | 1346 | |||
AK034138.1 | - | 5 | 1338 | |||
BC028916.1 | - | 5 | 1340 | 3676092 |