Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204171.1 | NP_001191100.1 | 7 | 620 | ||
NM_001278516.1 | NP_001265445.1 | 6 | 552 | |||
NM_001278517.1 | NP_001265446.1 | 4 | 326 | |||
NM_002393.4 | NP_002384.2 | 7 | 620 | |||
XM_017001311.1 | XP_016856800.1 | 7 | 619 | |||
XM_017001312.1 | XP_016856801.1 | 7 | 619 | |||
XM_017001313.1 | XP_016856802.1 | 7 | 619 | |||
GenBank mRNA | AF007111.1 | - | 7 | 570 | ||
AK223228.1 | - | 7 | 583 | |||
AK304698.1 | - | 6 | 525 | |||
AK310607.1 | - | 7 | 596 | |||
AK310903.1 | - | 6 | 525 | |||
AY949028.1 | - | 5 | 386 | |||
BC067299.1 | - | 7 | 610 | 30390159 | ||
BC105106.1 | - | 7 | 513 | 8144109 | ||
BC143431.1 | - | 6 | 445 | |||
BC143432.1 | - | 7 | 542 | |||
BX640923.1 | - | 7 | 565 | |||
CR749470.1 | - | 6 | 547 | |||
KC479043.1 | - | 6 | 454 | |||
KF435149.1 | - | 5 | 386 | |||
KF435150.1 | - | 5 | 386 |