Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204171.1 | NP_001191100.1 | 10 | 1127 | ||
NM_001204172.1 | NP_001191101.1 | 3 | 299 | |||
NM_001278516.1 | NP_001265445.1 | 10 | 1209 | |||
NM_001278517.1 | NP_001265446.1 | 8 | 983 | |||
NM_001278518.1 | NP_001265447.1 | 7 | 798 | |||
NM_001278519.1 | NP_001265448.1 | 5 | 608 | |||
NM_002393.4 | NP_002384.2 | 11 | 1277 | |||
XM_017001311.1 | XP_016856800.1 | 11 | 1276 | |||
XM_017001312.1 | XP_016856801.1 | 11 | 1261 | |||
XM_017001313.1 | XP_016856802.1 | 10 | 1126 | |||
GenBank mRNA | AB209383.1 | - | 2 | 2841 | ||
AF007111.1 | - | 11 | 1227 | |||
AK223228.1 | - | 11 | 1240 | |||
AK304698.1 | - | 9 | 1033 | |||
AK310607.1 | - | 11 | 1253 | |||
AY923176.1 | - | 3 | 187 | |||
AY949028.1 | - | 9 | 1043 | |||
BC067299.1 | - | 11 | 1267 | 30390159 | ||
BC105106.1 | - | 10 | 1020 | 8144109 | ||
BC143431.1 | - | 10 | 1102 | |||
BC143432.1 | - | 11 | 1199 | |||
BX640923.1 | - | 11 | 1222 |