Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026601.3 | NP_080877.3 | 4, 5 | 585 | ||
GenBank mRNA | AK004328.1 | - | 3, 4 | 173 | ||
AK020099.1 | - | 4, 5 | 459 | |||
BC024619.1 | - | 4, 5 | 573 | 4923664 | ||
BC100401.1 | - | 4, 5 | 549 | |||
CB952524.1 | - | 2, 3 | 90 | |||
CK620463.1 | - | 4, 5 | 473 |