Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MECOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004991.3 | NP_004982.2 | 2 | 524 | ||
XM_005247213.3 | XP_005247270.1 | 2 | 530 | |||
XM_005247214.3 | XP_005247271.1 | 2 | 530 | |||
XM_005247215.3 | XP_005247272.1 | 2 | 529 | |||
XM_005247224.3 | XP_005247281.1 | 2 | 530 | |||
XM_005247225.3 | XP_005247282.1 | 2 | 529 | |||
XM_005247226.3 | XP_005247283.1 | 2 | 407 | |||
XM_017005877.1 | XP_016861366.1 | 2 | 529 | |||
GenBank mRNA | AK304098.1 | - | 2 | 470 | ||
AK308404.1 | - | 2 | 169 | |||
AL700380.1 | - | 2 | 464 | |||
BC069498.1 | - | 2 | 373 | 7262213 | ||
CR541866.1 | - | 2 | 121 | |||
CR541886.1 | - | 2 | 121 | |||
DC347146.1 | - | 2 | 519 | |||
DC348381.1 | - | 2 | 510 | |||
U43292.1 | - | 2 | 428 | |||
U43293.1 | - | 1 | 84 |