Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leprotl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026609.2 | NP_080885.1 | 4 | 346 | ||
GenBank mRNA | AK003329.1 | - | 4 | 357 | ||
AK013870.1 | - | 4 | 416 | |||
AK075740.1 | - | 4 | 401 | |||
AK150289.1 | - | 4 | 405 | |||
AK153277.1 | - | 4 | 391 | |||
AK159401.1 | - | 4 | 334 | |||
AK203783.1 | - | 3 | 304 | |||
BC004677.1 | - | 4 | 346 | 3500223 |