Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaset2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026611.2 | NP_080887.1 | 4 | 396 | ||
GenBank mRNA | AK029149.1 | - | 350 | |||
AK084454.1 | - | 356 | ||||
BC100330.1 | - | 4 | 321 | 30527517 |
Gene Symbol: Rnaset2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083938.2 | NP_001077407.1 | 4 | 352 | ||
GenBank mRNA | AK014754.1 | - | 1 | 195 | ||
AK018722.1 | - | 4 | 352 | |||
AK019234.1 | - | 4 | 344 | |||
BC031496.1 | - | 4 | 332 | 3156549 | ||
BC089534.1 | - | 4 | 661 | 6703307 |