Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsto2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026619.2 | NP_080895.2 | 5 | 514 | ||
NM_030051.1 | NP_084327.1 | 5 | 467 | |||
GenBank mRNA | AK018953.1 | - | 5 | 506 | ||
AK019582.1 | - | 5 | 433 | |||
AK075753.1 | - | 5 | 515 | |||
AK077086.1 | - | 5 | 468 | |||
BC030371.1 | - | 3 | 372 | 5371983 |