Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AI846148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033139.3 | NP_001028311.2 | 6 | 1317 | ||
GenBank mRNA | AK090385.1 | - | 6 | 1286 | ||
AK142757.1 | - | 5 | 713 | |||
AK163590.1 | - | 6 | 1118 | |||
AK166227.1 | - | 6 | 1287 | |||
BC125249.1 | - | 6 | 1145 | 40059530 |