Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A930018P22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026634.2 | NP_080910.1 | 2 | 696 | ||
GenBank mRNA | AK006944.1 | - | 2 | 690 | ||
AK020874.1 | - | 2 | 697 | |||
BC048438.1 | - | 2 | 655 | 6706651 |