Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177576.1 | NP_001171047.1 | 5 | 502 | ||
NM_026646.3 | NP_080922.1 | 5 | 744 | |||
GenBank mRNA | AK010193.1 | - | 5 | 490 | ||
AK046874.1 | - | 5 | 489 | |||
AK048389.1 | - | 5 | 473 | |||
AK049313.1 | - | 5 | 473 | |||
AK155276.1 | - | 5 | 496 | |||
AK162400.1 | - | 5 | 455 | |||
BC037949.1 | - | 5 | 501 | 5367108 |