Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomgnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290658.1 | NP_001277587.1 | 16 | 1595 | ||
NM_026651.2 | NP_080927.1 | 17 | 1681 | |||
NM_029786.2 | NP_084062.2 | 16 | 1582 | |||
GenBank mRNA | AB053220.1 | - | 17 | 1663 | ||
AK002638.1 | - | 16 | 1574 | |||
AK015478.1 | - | 16 | 1597 | |||
AK019640.1 | - | 16 | 1587 | |||
AK178221.1 | - | 1 | 45 | |||
AK180366.1 | - | 1 | 45 | |||
AK210391.1 | - | 1 | 45 | |||
AK212205.1 | - | 1 | 45 | |||
BC011201.1 | - | 17 | 1604 | 4190368 | ||
BC023265.1 | - | 14 | 1269 |