Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930430F08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175128.2 | NP_780337.2 | 5 | 622 | ||
GenBank mRNA | AK015251.1 | - | 5 | 571 | ||
AK016270.1 | - | 454 | ||||
AK032226.1 | - | 5 | 578 | |||
AK036647.1 | - | 5 | 605 | |||
AK077761.1 | - | 5 | 617 | |||
AK078231.1 | - | 5 | 623 | |||
AK082894.1 | - | 5 | 575 | |||
AK160693.1 | - | 5 | 605 | |||
BC082587.1 | - | 5 | 572 | 30630155 |