Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024222.2 | NP_077184.2 | 12 | 2179 | ||
GenBank mRNA | AK012153.1 | - | 12 | 1849 | ||
AK018758.2 | - | 12 | 1889 | |||
AK145674.1 | - | 12 | 1820 | |||
AK148065.1 | - | 12 | 1878 | |||
AK152899.1 | - | 12 | 2227 | |||
AK154979.1 | - | 12 | 1868 | |||
AK170079.1 | - | 12 | 1891 | |||
AK181242.1 | - | 1 | 23 | |||
AK205163.1 | - | 1 | 26 | |||
AX799084.1 | - | 12 | 1890 | |||
BC003206.1 | - | 7 | 891 | |||
BC013054.1 | - | 2 | 297 | |||
BC052433.1 | - | 12 | 2179 | 6837097 |