Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024222.2 | NP_077184.2 | 15 | 2744 | ||
GenBank mRNA | AK012153.1 | - | 15 | 2414 | ||
AK013164.1 | - | 1 | 28 | |||
AK017675.1 | - | 1 | 28 | |||
AK018758.2 | - | 15 | 2454 | |||
AK145674.1 | - | 15 | 2385 | |||
AK148065.1 | - | 15 | 2443 | |||
AK152899.1 | - | 15 | 2792 | |||
AK154979.1 | - | 15 | 2433 | |||
AK170079.1 | - | 15 | 2456 | |||
AX799084.1 | - | 15 | 2455 | |||
BC003206.1 | - | 10 | 1456 | |||
BC013054.1 | - | 5 | 862 | |||
BC052433.1 | - | 15 | 2744 | 6837097 |