Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146171.1 | NP_666283.1 | 10 | 1282 | ||
GenBank mRNA | AK013380.1 | - | 1 | 96 | ||
AK129068.1 | - | 10 | 1247 | |||
AK156143.1 | - | 10 | 1312 | |||
BC006673.1 | - | 3 | 428 | |||
BC008592.1 | - | 1 | 176 | |||
BC025460.1 | - | 10 | 1302 | |||
BC026982.1 | - | 10 | 1282 | 5364933 | ||
BC029133.1 | - | 10 | 1239 | 4951596 |