Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam114a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026667.3 | NP_080943.3 | 6 | 831 | ||
GenBank mRNA | AK007950.1 | - | 6 | 789 | ||
AK014725.1 | - | 6 | 801 | |||
AK017336.1 | - | 6 | 791 | |||
AK020262.1 | - | 6 | 792 | |||
AK030360.1 | - | 6 | 832 | |||
AM162548.1 | - | 6 | 835 | |||
BC027235.1 | - | 6 | 794 | 3603083 | ||
BC110359.1 | - | 6 | 832 | 5325099 |