Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam114a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026667.3 | NP_080943.3 | 12 | 1578 | ||
GenBank mRNA | AK007950.1 | - | 12 | 1536 | ||
AK017336.1 | - | 12 | 1538 | |||
AK020262.1 | - | 12 | 1539 | |||
AK030360.1 | - | 12 | 1579 | |||
AM162548.1 | - | 12 | 1582 | |||
BC027235.1 | - | 12 | 1541 | 3603083 | ||
BC110359.1 | - | 12 | 1579 | 5325099 |