Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam114a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026667.3 | NP_080943.3 | 6 | 802 | ||
GenBank mRNA | AK007950.1 | - | 6 | 760 | ||
AK014725.1 | - | 6 | 772 | |||
AK017336.1 | - | 6 | 762 | |||
AK020262.1 | - | 6 | 763 | |||
AK030360.1 | - | 6 | 803 | |||
AM162548.1 | - | 6 | 806 | |||
BC027235.1 | - | 6 | 765 | 3603083 | ||
BC110359.1 | - | 6 | 803 | 5325099 |