Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026671.1 | NP_080947.1 | 3 | 311 | ||
GenBank mRNA | AB009019.1 | - | 3 | 327 | ||
AK002226.1 | - | 3 | 327 | |||
BC099511.1 | - | 3 | 311 | 30845404 | ||
BC132407.1 | - | 3 | 312 | 40130684 | ||
BC132409.1 | - | 3 | 312 | 40130686 |