Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026672.2 | NP_080948.2 | 6 | 500 | ||
GenBank mRNA | AK002213.1 | - | 6 | 473 | ||
AK162441.1 | - | 6 | 423 | |||
BC051924.1 | - | 6 | 500 | 5692787 |