Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026675.2 | NP_080951.1 | 2 | 232 | ||
GenBank mRNA | AK002252.1 | - | 2 | 233 | ||
BC019768.1 | - | 1 | 209 | 5124658 |