Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEF2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271629.1 | NP_001258558.1 | 11 | 2207 | ||
NM_005920.3 | NP_005911.1 | 12 | 2509 | |||
XM_005245169.4 | XP_005245226.1 | 12 | 2491 | |||
XM_005245170.3 | XP_005245227.1 | 12 | 2516 | |||
XM_006711330.3 | XP_006711393.1 | 12 | 2220 | |||
XM_006711332.3 | XP_006711395.1 | 12 | 2504 | |||
XM_006711333.2 | XP_006711396.1 | 11 | 2483 | |||
XM_006711334.3 | XP_006711397.1 | 11 | 2481 | |||
XM_011509569.2 | XP_011507871.1 | 12 | 2541 | |||
XM_017001314.1 | XP_016856803.1 | 11 | 2625 | |||
XM_017001315.1 | XP_016856804.1 | 11 | 2220 | |||
GenBank mRNA | AB210005.1 | - | 11 | 2759 | ||
AK056972.1 | - | 1 | 220 | |||
BC040949.1 | - | 11 | 2207 | 6044434 | ||
BC054520.1 | - | 12 | 2404 | 6142751 | ||
BX647226.1 | - | 1 | 246 |