Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026680.4 | NP_080956.1 | 2 | 146 | ||
GenBank mRNA | AK002581.1 | - | 2 | 154 | ||
AK141378.1 | - | 2 | 147 | |||
AY707855.1 | - | 2 | 169 | |||
BC024448.1 | - | 2 | 169 | 4974299 |