Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc88c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026681.4 | NP_080957.2 | 30 | 5493 | ||
GenBank mRNA | AK173188.1 | - | 17 | 3307 | ||
AY339881.1 | - | 30 | 5494 | |||
BC010253.1 | - | 1 | 100 | |||
BC043115.1 | - | 29 | 4599 | 6408630 | ||
BC063255.1 | - | 29 | 4599 | 6408630 |