Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026684.2 | NP_080960.1 | 1, 2 | 208 | ||
GenBank mRNA | AK002517.1 | - | 1, 2 | 172 | ||
AK151404.1 | - | 1, 2 | 242 | |||
BC031664.1 | - | 1, 2 | 208 | 4219237 | ||
BC043013.1 | - | 1, 2 | 185 | 5683280 |