Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026697.3 | NP_080973.1 | 8 | 887 | ||
GenBank mRNA | AB232609.1 | - | 7 | 518 | ||
AJ968955.1 | - | 8 | 742 | |||
AJ968956.1 | - | 7 | 688 | |||
AJ968957.1 | - | 8 | 742 | |||
AJ968958.1 | - | 7 | 688 | |||
AJ968959.1 | - | 8 | 742 | |||
AJ968960.1 | - | 7 | 688 | |||
AK002704.1 | - | 8 | 759 | |||
AK133963.1 | - | 8 | 787 | |||
AK135751.1 | - | 8 | 777 | |||
AK160859.1 | - | 8 | 887 | |||
AK165812.1 | - | 8 | 734 | |||
AK166863.1 | - | 1 | 906 | |||
AK168551.1 | - | 8 | 744 | |||
AK168867.1 | - | 8 | 756 | |||
AK169396.1 | - | 8 | 758 | |||
AK216803.1 | - | 5 | 305 | |||
BC009085.1 | - | 8 | 570 | 2649852 | ||
BC025139.1 | - | 8 | 758 | 3980228 | ||
BC056648.1 | - | 8 | 760 | 5034519 |